siRNA Details
Gene Symbol: 9430083A17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_029463.1 | 1 | 1611 | |||
GenBank mRNA | AK083743.1 | - | 1 | 1579 | ||
AK157883.1 | - | 1 | 1604 | |||
AK158202.1 | - | 1 | 1605 |
