siRNA Details
Gene Symbol: 9430083A17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_029463.1 | 1 | 1115 | |||
GenBank mRNA | AK020502.1 | - | 1 | 437 | ||
AK083743.1 | - | 1 | 1083 | |||
AK157883.1 | - | 1 | 1107 | |||
AK158202.1 | - | 1 | 1106 |
