siRNA Details
Gene Symbol: 9430083A17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_029463.1 | 1 | 1336 | |||
GenBank mRNA | AK020502.1 | - | 1 | 658 | ||
AK083743.1 | - | 1 | 1304 | |||
AK157883.1 | - | 1 | 1329 | |||
AK158202.1 | - | 1 | 1327 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BI466564.1 | - | 1 | 186 | Chr. 13 - 51099288 - 51099559 [-] on Build GRCm38 |
