siRNA Details
Gene Symbol: Mir22hg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
RefSeq | NR_030711.1 | 3 | 869 | |||
GenBank mRNA | AK008813.1 | - | LIT3441 2210403K04Rik |
3 | 761 | |
AK018628.1 | - | 3 | 779 | |||
AK021103.1 | - | 3 | 870 | |||
BG076757.2 | - | 1 | 163 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BF786183.1 | - | 530 | Chr. - - [-] on Build |
