siRNA Details
Gene Symbol: Mir22hg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
RefSeq | NR_030711.1 | 3 | 1643 | |||
GenBank mRNA | AK008813.1 | - | LIT3441 2210403K04Rik |
3 | 1535 | |
AK018628.1 | - | 3 | 1553 |
