siRNA Details
Gene Symbol: Atp2a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027838.1 | 20 | 4538 | |||
GenBank mRNA | AK081191.1 | - | 1 | 175 | ||
AK134659.1 | - | 1 | 707 | |||
AK220538.1 | - | 20 | 4487 | |||
BC054531.1 | - | 20 | 4197 | 6406916 | ||
BC054748.1 | - | 20 | 4197 | 6406916 |
