siRNA Details
Gene Symbol: Gm8883 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027658.1 | 3 | 1003 | |||
NR_027659.1 | 3 | 683 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY735181.1 | - | 3 | 681 | Chr. 1 - 71888112 - 71891200 [+] on Build GRCm38 |
