siRNA Details
Gene Symbol: AI854703 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027236.1 | 1 | 67 | |||
GenBank mRNA | AK038960.1 | - | 1 | 300 | ||
BC145738.1 | - | 1 | 67 | |||
BC145740.1 | - | 1 | 67 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: AI854703 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027236.1 | 1 | 67 | |||
GenBank mRNA | AK038960.1 | - | 1 | 300 | ||
BC145738.1 | - | 1 | 67 | |||
BC145740.1 | - | 1 | 67 |