siRNA Details
Gene Symbol: 2900005J15Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027851.1 | 1 | 675 | |||
GenBank mRNA | AK013483.1 | - | 1 | 676 | ||
AK015504.1 | - | 192 | ||||
AK076604.1 | - | 1 | 668 | |||
BC094288.1 | - | 1 | 710 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | NM_026059.1 | - | 668 | Chr. - - [-] on Build |
