siRNA Details
Gene Symbol: Snora41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028558.1 | 1 | 43 | |||
GenBank mRNA | AF357411.1 | - | 1 | 43 |

More Information
Other Species Targeted

Gene Ontology Categories:
