siRNA Details
Gene Symbol: 1700012D14Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015573.2 | 3 | 595 | |||
GenBank mRNA | AK005898.1 | - | 3 | 597 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AV040400.2 | - | 2 | 289 | Chr. 7 - 111117673 - 111121010 [-] on Build GRCm38 |
