siRNA Details
Gene Symbol: Gm5126 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026596.1 | 1 | 2000 | |||
GenBank mRNA | AK077703.1 | - | 1 | 2001 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gm5126 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026596.1 | 1 | 2000 | |||
GenBank mRNA | AK077703.1 | - | 1 | 2001 |