siRNA Details
Gene Symbol: Vmn2r-ps11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003962.1 | 1 | 86 | |||
GenBank mRNA | AK029734.1 | - | 1 | 87 | ||
BB615497.1 | - | 1 | 87 |
Gene Symbol: Vmn2r7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK030224.1 | - | 108 |
