siRNA Details
Gene Symbol: Rmst | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028262.1 | 9 | 895 | |||
GenBank mRNA | AK086758.1 | - | 9 | 888 | ||
AK136927.1 | - | 5 | 1725 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BQ572455.1 | - | 6 | 551 | Chr. 10 - 92083887 - 92164663 [-] on Build GRCm38 |
