siRNA Details
Gene Symbol: Rmst | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028262.1 | 7 | 670 | |||
GenBank mRNA | AK086758.1 | - | 7 | 663 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CO425135.1 | - | 6 | 568 | Chr. 10 - 92133279 - 92164739 [-] on Build GRCm38 |
