siRNA Details
Gene Symbol: 1700047M11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015458.1 | 2 | 613 | |||
GenBank mRNA | AK006718.1 | - | 2 | 397 | ||
AK019698.1 | - | 2 | 392 | |||
AK047804.1 | - | 2374 | ||||
AV345418.1 | - | 1 | 164 | |||
BC048591.1 | - | 2 | 613 |
