siRNA Details
Gene Symbol: 1700047M11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015458.1 | 1 | 244 | |||
GenBank mRNA | AK006718.1 | - | 1 | 28 | ||
AK019698.1 | - | 1 | 23 | |||
BC048591.1 | - | 1 | 244 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY715448.1 | - | 1 | 23 | Chr. 1 - 182301056 - 182303248 [+] on Build GRCm38 |
