siRNA Details
Gene Symbol: 3830408C21Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015471.1 | 4 | 1670 | |||
GenBank mRNA | AK014423.1 | - | 4 | 1670 | ||
AK043130.1 | - | 5 | 1459 | |||
AK052571.1 | - | 1 | 1329 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB253732.2 | - | 2 | 78 | Chr. 13 - 107032898 - 107033521 [+] on Build GRCm38 |
