siRNA Details
Gene Symbol: 5530601H04Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015467.1 | 5 | 1968 | |||
GenBank mRNA | AK084653.1 | - | 6 | 2043 | ||
AK084980.1 | - | 6 | 2022 | |||
AK085098.1 | - | 6 | 2014 | |||
AK153718.1 | - | 771 | ||||
AK164650.1 | - | 5 | 1969 |
