siRNA Details
Gene Symbol: 5530601H04Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015467.1 | 5 | 519 | |||
GenBank mRNA | AK017459.1 | - | 456 | |||
AK043766.1 | - | 594 | ||||
AK080296.1 | - | 6 | 589 | |||
AK084653.1 | - | 6 | 594 | |||
AK084980.1 | - | 6 | 573 | |||
AK085098.1 | - | 6 | 566 | |||
AK085485.1 | - | 6 | 560 | |||
AK153718.1 | - | 331 | ||||
AK157237.1 | - | 552 | ||||
AK164650.1 | - | 5 | 520 | |||
BC019699.1 | - | 334 | ||||
BC049927.1 | - | 308 | ||||
BC055463.1 | - | 6 | 468 |
