siRNA Details
Gene Symbol: 1700084E18Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028299.1 | 1 | 211 | |||
GenBank mRNA | AK006990.1 | - | 1 | 212 | ||
AK018989.1 | - | 1 | 100 | |||
AK018997.1 | - | 1 | 100 | |||
AK034708.1 | - | 805 | ||||
BC062268.1 | - | 1 | 187 |
