siRNA Details
Gene Symbol: Snora16a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_029412.1 | 1 | 65 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CA977667.1 | - | 1 | 183 | Chr. 4 - 132309347 - 132310223 [+] on Build GRCm38 |
