siRNA Details
Gene Symbol: Il1bos | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015474.1 | 3 | 660 | |||
GenBank mRNA | AK035070.1 | - | 3 | 661 | ||
AK076405.1 | - | 4 | 1675 | |||
BC111869.1 | - | 1 | 1871 | |||
BC112374.1 | - | 1 | 1869 |
