siRNA Details
Gene Symbol: Il1bos | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015474.1 | 2 | 373 | |||
GenBank mRNA | AK035070.1 | - | 2 | 374 | ||
AK076405.1 | - | 3 | 232 | |||
BC111869.1 | - | 1 | 20 | |||
BC112374.1 | - | 1 | 18 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB627324.1 | - | 2 | 374 | Chr. 2 - 129369902 - 129374957 [+] on Build GRCm38 |
