siRNA Details
Gene Symbol: Il1bos | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015474.1 | 3 | 1088 | |||
GenBank mRNA | AK035070.1 | - | 3 | 1089 | ||
AK076405.1 | - | 4 | 2103 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Il1bos | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015474.1 | 3 | 1088 | |||
GenBank mRNA | AK035070.1 | - | 3 | 1089 | ||
AK076405.1 | - | 4 | 2103 |