siRNA Details
Gene Symbol: 9130230L23Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027961.1 | 7 | 1127 | |||
GenBank mRNA | AK033717.1 | - | 7 | 1128 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 9130230L23Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027961.1 | 7 | 1127 | |||
GenBank mRNA | AK033717.1 | - | 7 | 1128 |