siRNA Details
Gene Symbol: 9130230L23Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027961.1 | 4 | 437 | |||
GenBank mRNA | AK033717.1 | - | 4 | 438 | ||
BC096478.1 | - | 4 | 540 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 9130230L23Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027961.1 | 4 | 437 | |||
GenBank mRNA | AK033717.1 | - | 4 | 438 | ||
BC096478.1 | - | 4 | 540 |