siRNA Details
Gene Symbol: 9130230L23Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027961.1 | 2 | 187 | |||
GenBank mRNA | AK033717.1 | - | 2 | 188 | ||
AK166951.1 | - | 1 | 463 | |||
BC096478.1 | - | 2 | 176 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 9130230L23Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027961.1 | 2 | 187 | |||
GenBank mRNA | AK033717.1 | - | 2 | 188 | ||
AK166951.1 | - | 1 | 463 | |||
BC096478.1 | - | 2 | 176 |