siRNA Details
Gene Symbol: 4921511C20Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003646.1 | 1 | 1403 | |||
GenBank mRNA | AK076563.1 | - | 1 | 1403 | ||
BC125512.1 | - | 1 | 1362 | |||
BC132192.1 | - | 1 | 1362 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AV254564.2 | - | 1 | 459 | Chr. X - 127395238 - 127395898 [+] on Build GRCm38 |
