siRNA Details
Gene Symbol: 4921511C20Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003646.1 | 1 | 744 | |||
GenBank mRNA | AK076563.1 | - | 1 | 744 | ||
BC125512.1 | - | 1 | 703 | |||
BC132192.1 | - | 1 | 703 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 4921511C20Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003646.1 | 1 | 744 | |||
GenBank mRNA | AK076563.1 | - | 1 | 744 | ||
BC125512.1 | - | 1 | 703 | |||
BC132192.1 | - | 1 | 703 |