siRNA Details
Gene Symbol: 8430429K09Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028317.1 | 1 | 189 | |||
NR_045276.1 | 1 | 143 | ||||
GenBank mRNA | AK086952.1 | - | 1 | 154 | ||
BY069744.1 | - | 1 | 144 | |||
BY284012.1 | - | 1 | 192 | |||
BY331368.1 | - | 1 | 146 |
