siRNA Details
Gene Symbol: Gm12216 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033332.1 | 1 | 14 | |||
GenBank mRNA | AK138811.1 | - | 1 | 15 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY724574.1 | - | 1 | 15 | Chr. 11 - 53786005 - 53859256 [-] on Build GRCm38 |
