siRNA Details
Gene Symbol: Gm12216 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033332.1 | 3 | 277 | |||
GenBank mRNA | AK039036.1 | - | 2 | 101 | ||
AK138811.1 | - | 3 | 278 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gm12216 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033332.1 | 3 | 277 | |||
GenBank mRNA | AK039036.1 | - | 2 | 101 | ||
AK138811.1 | - | 3 | 278 |