siRNA Details
Gene Symbol: 1700030C10Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015521.1 | 2 | 1310 | |||
GenBank mRNA | AK006529.1 | - | 2 | 1098 | ||
BC100365.1 | - | 2 | 1310 |
Gene Symbol: Gm9222 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | CA748277.1 | - | 1 | 133 |
