siRNA Details
Gene Symbol: Gm5577 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026990.1 | 2 | 569 | |||
GenBank mRNA | AK042699.1 | - | 582 | |||
AK048951.1 | - | 2 | 541 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB650272.1 | - | 2 | 541 | Chr. 6 - 87981712 - 87983439 [+] on Build GRCm38 |
