siRNA Details
Gene Symbol: Tug1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002321.2 | 3 | 705 | |||
NR_002322.2 | 2 | 1101 | ||||
NR_110490.1 | 2 | 1101 | ||||
GenBank mRNA | AK033381.1 | - | 2 | 1073 | ||
AK048213.1 | - | 2 | 1093 | |||
AK083955.1 | - | 2 | 1084 | |||
AK088174.1 | - | 3 | 706 | |||
BC016239.1 | - | 2 | 450 | |||
BC048828.1 | - | 2 | 977 | |||
DQ517434.1 | - | 2 | 1102 | |||
DQ517435.1 | - | 3 | 701 |
