siRNA Details
Gene Symbol: Tug1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002321.2 | 3 | 1180 | |||
NR_002322.2 | 2 | 1576 | ||||
NR_110490.1 | 2 | 1576 | ||||
GenBank mRNA | AK033381.1 | - | 2 | 1548 | ||
AK048213.1 | - | 2 | 1568 | |||
AK083955.1 | - | 2 | 1560 | |||
AK088174.1 | - | 3 | 1181 | |||
BC016239.1 | - | 2 | 925 | |||
BC048828.1 | - | 2 | 1452 | |||
DQ517434.1 | - | 2 | 1577 | |||
DQ517435.1 | - | 3 | 1176 |
