siRNA Details
Gene Symbol: 5730408K05Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027866.1 | 1 | 83 | |||
GenBank mRNA | AK017527.1 | - | 84 | |||
AK019951.1 | - | 1 | 78 | |||
AK043270.1 | - | 1 | 84 | |||
BC027679.1 | - | 1 | 69 |
