siRNA Details
Gene Symbol: A930007I19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015567.2 | 4 | 1894 | |||
GenBank mRNA | AK137192.1 | - | 4 | 1896 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: A930007I19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015567.2 | 4 | 1894 | |||
GenBank mRNA | AK137192.1 | - | 4 | 1896 |