siRNA Details
Gene Symbol: Gm5627 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033301.1 | 4 | 916 | |||
GenBank mRNA | AK041874.1 | - | 4 | 919 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB233413.2 | - | 3 | 653 | Chr. 9 - 102747519 - 102756684 [+] on Build GRCm38 |
