siRNA Details
Gene Symbol: Gm5627 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033301.1 | 1 | 213 | |||
GenBank mRNA | AK041874.1 | - | 1 | 216 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB638603.1 | - | 1 | 215 | Chr. 9 - 102739648 - 102749374 [+] on Build GRCm38 |
