siRNA Details
Gene Symbol: Gm5627 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033301.1 | 4 | 627 | |||
GenBank mRNA | AK041874.1 | - | 4 | 630 | ||
BC147467.1 | - | 4 | 381 | 9056109 | ||
BC147470.1 | - | 4 | 381 | 9056112 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB233413.2 | - | 3 | 364 | Chr. 9 - 102747519 - 102756684 [+] on Build GRCm38 |
