siRNA Details
Gene Symbol: Gm5627 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033301.1 | 3 | 540 | |||
GenBank mRNA | AK041874.1 | - | 3 | 543 | ||
BC147467.1 | - | 3 | 294 | 9056109 | ||
BC147470.1 | - | 3 | 294 | 9056112 |
