siRNA Details
Gene Symbol: BC024386 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015583.1 | 1 | 228 | |||
GenBank mRNA | BC014806.1 | - | 1 | 244 | ||
BC039766.1 | - | 1 | 248 | |||
BC087948.1 | - | 1 | 228 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: BC024386 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015583.1 | 1 | 228 | |||
GenBank mRNA | BC014806.1 | - | 1 | 244 | ||
BC039766.1 | - | 1 | 248 | |||
BC087948.1 | - | 1 | 228 |