siRNA Details
Gene Symbol: BC024386 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015583.1 | 1 | 34 | |||
GenBank mRNA | BC014806.1 | - | 1 | 50 | ||
BC024386.1 | - | 1 | 56 | |||
BC039766.1 | - | 1 | 54 | |||
BC061156.1 | - | 1 | 15 | |||
BC087948.1 | - | 1 | 34 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BI147306.1 | - | 1 | 55 | Chr. 7 - 140906043 - 140907979 [+] on Build GRCm38 |
