siRNA Details
Gene Symbol: BC024386 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015583.1 | 1 | 327 | |||
GenBank mRNA | BC014806.1 | - | 1 | 343 | ||
BC039766.1 | - | 1 | 347 | |||
BC087948.1 | - | 1 | 327 | |||
BC100589.1 | - | 2 | 77 |
