siRNA Details
Gene Symbol: BC024386 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015583.1 | 2 | 1338 | |||
GenBank mRNA | BC014806.1 | - | 2 | 1354 | ||
BC024386.1 | - | 4 | 449 | |||
BC039766.1 | - | 2 | 1358 | |||
BC061156.1 | - | 5 | 612 | |||
BC087948.1 | - | 2 | 1338 | |||
BC100589.1 | - | 4 | 488 |
