siRNA Details
Gene Symbol: A930015D03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015618.2 | 2 | 616 | |||
GenBank mRNA | AK020860.1 | - | 2 | 617 | ||
AK078380.1 | - | 2 | 666 | |||
AK142942.1 | - | 2 | 612 | |||
BC055918.1 | - | 2 | 594 |
Gene Symbol: BC023719 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC023719.1 | - | 2 | 537 |
