siRNA Details
Gene Symbol: A930015D03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015618.2 | 2 | 509 | |||
GenBank mRNA | AK020860.1 | - | 2 | 510 | ||
AK078380.1 | - | 2 | 560 | |||
AK142942.1 | - | 2 | 505 | |||
BC055918.1 | - | 2 | 487 |
