siRNA Details
Gene Symbol: A930015D03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015618.2 | 1, 2 | 64 | |||
GenBank mRNA | AK020860.1 | - | 1, 2 | 65 | ||
AK078380.1 | - | 1, 2 | 115 | |||
AK142942.1 | - | 1, 2 | 60 | |||
BC055918.1 | - | 1, 2 | 42 |
